Anti-FAM13B1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Toimittaja: Bioss
BOSSBS-5757R-FITCEA 858 EUR
BOSSBS-5757R-FITC
Anti-FAM13B1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
FAM13B1

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM13B1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR