Anti-TMED4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Toimittaja: Bioss
BOSSBS-6208R-FITCEA 858 EUR
BOSSBS-6208R-FITC
Anti-TMED4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
TMED4 is a putative NF-kB activating protein belonging to the EMP24/GP25L family, and contains 1 GOLD domain. It is also known as Transmembrane emp24 protein transport domain containing 4.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMED4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR