Knitted gloves made from Dyneema®/glass fibre/polyamide. Rough black nitrile coating on palm and fingers. Ventilated back. The Dyneema®/glass-fibre yarn offers exceptional resistance against cuts (level 5) and abrasion (level 4). The nitrile coating means that the gloves can be used for work involving contact with oil and offers significantly improved mechanical resistance.
- The knit design ensures a close-fitting glove with excellent tactile sensitivity, maximum comfort and a high level of resistance.
- The Dyneema® fibre ensures comfort in contact with the hand.
- Dyneema® is resistant to most chemicals (detergents), which makes the gloves easier to wash.
Length: 260 mm
Gauge: 10 (10 stiches per inch)
Colour: Green and white blend
EN 420, EN 388