VWR® Hitachi NOBIAS, Solid phase packed columns, chelate-PA1
Toimittaja: Avantor
HITA890-5377EA 4030 EUR
HITA890-5377
VWR® Hitachi NOBIAS, Solid phase packed columns, chelate-PA1
Kromatografiakolonnit
NOBIAS series are designed for the pre-treatment method to divide the foreign matter and analytical target compound. Used for concentrating transition metals in water samples.
  • High liquid permeability
  • Improved measurement accuracy
  • Reduction of preprocessing time
  • Optimise packing materials by application
  • Adoption of new filler