Anti-PXN Rabbit Monoclonal Antibody [clone: RM256]

Toimittaja: Abnova
ABNOMAB15135EA 440 EUR
ABNOMAB15135
Anti-PXN Rabbit Monoclonal Antibody [clone: RM256]
Vasta-aineet
Rabbit monoclonal antibody raised against synthetic peptide of human PXN.

Type: Primary
Antigen: PXN
Clonality: Monoclonal
Clone: RM256
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR