Anti-CD206 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB180464-100EA 0 EUR
BIRBORB180464-100
Anti-CD206 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CD206
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR