HEPES -natriumsuola ≥99.0% (dried basis), puhdistettu, J.T. Baker®
Toimittaja: Avantor
4809-02EA
1138.41
EUR
4809-02
4809-06
4809-04
HEPES -natriumsuola ≥99.0% (dried basis), puhdistettu, J.T. Baker®
HEPES -natriumsuola
Suitable for use in biopharmaceutical manufacturing applications.
Kaava:
C₈H₁₇N₂NaO₄S MW: 260,29 g/mol Sulamispiste: 105 °C Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00036463 CAS-numero: 75277-39-3 EINECS: 278-169-7 |
![]() |
Testitulosten erittely
For Laboratory,Research,or Manufacturing Use | |
Assay (C₈H₁₇N₂NaO₄S) (dried basis) | ≥99.0 % |
Appearance | Passes Test |
Solubility | Passes Test |
Loss on Drying at 80°C | ≤3.0 % |
Insoluble Matter | ≤1.0 % |
Heavy Metals (as Pb) | ≤1 ppm |
Identification (Sodium) | Passes Test |
DNase Activity (None Detected) | Passes Test |
RNase Activity | Passes Test |
Protease Activity | Passes Test |
pK^a at 20°C, For Information Only | |
QC Supervisor or equivalent to check supplier's COA to ensure proper chemical form. | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...