Mangaani(II)karbonaatti Mn ≥ 43,0%, jauhe, BAKER ANALYZED®, Laboratory Reagent, J.T. Baker®
Toimittaja: Avantor
2536-07EA
865.57
EUR
2536-07
2536-R
Mangaani(II)karbonaatti Mn ≥ 43,0%, jauhe, BAKER ANALYZED®, Laboratory Reagent, J.T. Baker®
Mangaani(II)karbonaatti
Powder. Lot analysis on label.
Kaava:
CMnO₃ MW: 114,95 g/mol Tiheys: 3,12 g/cm³ (20 °C) |
MDL numero:
MFCD00011116 CAS-numero: 598-62-9 EINECS: 209-942-9 Merck Index: 12,05766 |
Testitulosten erittely
For Laboratory,Research,or Manufacturing Use | |
Assay (as Mn) | ≥ 43.0 % |
Insoluble in HCl | ≤ 0.01 % |
Calcium (Ca) | ≤ 0.01 % |
Chloride (Cl) | ≤ 0.02 % |
Magnesium (Mg) | ≤ 0.01 % |
Potassium (K) | ≤ 0.01 % |
Sulfate (SO₄) | ≤ 0.005 % |
Other Heavy Metals (as Pb) | ≤ 0.005 % |
Iron (Fe) | ≤ 0.02 % |
Sodium (Na) | ≤ 0.02 % |
Zinc (Zn) | ≤ 0.05 % |
Average Particle Diameter, µm (APD) | ≤ 3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...