Natriumtiosyanaatti liuos, BAKER ANALYZED®, J.T.Baker®
Toimittaja: Avantor
|
Varoitus
|
3939-09EA
0
EUR
3939-09
Natriumtiosyanaatti liuos, BAKER ANALYZED®, J.T.Baker®
Natriumtiosyanaatti
Kaava:
NaSCN MW: 81,07 g/mol |
MDL numero:
MFCD00011123 CAS-numero: 540-72-7 |
Testitulosten erittely
For Laboratory,Research,or Manufacturing Use | |
Density (g/mL) at 20°C | 1.3064 - 1.3122 |
Insoluble Matter | ≤0.002 % |
Carbonate (as Na₂CO₃) | ≤0.1 % |
Chloride (Cl) | ≤0.002 % |
Sulfate (SO₄) | ≤0.005 % |
Sulfide (S) | ≤0.0005 % |
Ammonium (NH₄) | ≤0.001 % |
Reducing Agents (x10⁻⁵ meq/gm), For Information Only | |
Heavy Metals (as Pb) | ≤2 ppm |
Trace Impurities - Iron (Fe) | ≤1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...