Labels, tags, blank utility tags
Toimittaja: Brady
BRDY087699EA 3.8 EUR
BRDY087699 BRDY041922 BRDY041921 BRDY041924 BRDY041923 BRDY041926 BRDY087695 BRDY041925 BRDY087696 BRDY087697 BRDY041928 BRDY041927 BRDY087698 BRDY041929 BRDY087694 BRDY041920
Labels, tags, blank utility tags
Etiketit, tarrat
These durable tags are made from laminated polyester on a plastic tag with rounded corners and a punched hole.
  • Lightweight
  • Non corrosive, non conductive and impact resistant
  • Label area that allows your own custom printed label to be attached
Blank tags allow users to enter the desired wording on the floor or at any time. Tags may be completely blank, or come with a header (eg. 'Warning' or 'Notice'). Signed by/Date information on the bottom should be captured as text legend.