Anti-NUP50 Goat Polyclonal Antibody

Toimittaja: ProSci Inc.
PRSI45-985EA 553 EUR
PRSI45-985
Anti-NUP50 Goat Polyclonal Antibody
Vasta-aineet
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 60 kDa band observed in Jurkat lysate (calculated MW 50.1 kDa according to NP_009103.2). Recommended for use at 0.2-1 μg/mL. Immunohistochemistry: In paraffin embedded human uterus shows textured nuclear staining of endometrial cells. Recommended concentration, 2.5-3.8 μg/mL.

Nup50 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Nup50 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NUP50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR