Anti-POU3F1 Goat Polyclonal Antibody

Toimittaja: ProSci Inc.
PRSI45-992EA 435 EUR
PRSI45-992
Anti-POU3F1 Goat Polyclonal Antibody
Vasta-aineet
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Preliminary experiments in lysates of human brain (cerebellum, hippocampus, frontal cortex), mouse brain, rat brain and rat spinal cord gave no specific signal but low background (at antibody concentration up to 1 μg/mL).

OCT6 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

OCT6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: POU3F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR