Anti-OGFR Goat Polyclonal Antibody

Toimittaja: ProSci Inc.
PRSI45-993EA 494 EUR
PRSI45-993
Anti-OGFR Goat Polyclonal Antibody
Vasta-aineet
ELISA: Antibody detection limit dilution 1:4,000.

OGFR antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

OGFR antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: OGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR