ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 50 kDa band observed in human brain (cerebral cortex, hippocampus and substantia nigra) lysates (calculated MW of 50.7 kDa according to NP_001517.1). Recommended concentration: 1-3 μg/mL.
Orexin Receptor 2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Orexin Receptor 2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HCRTR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human