Anti-OSBPL1A Goat Polyclonal Antibody
Toimittaja: ProSci Inc.
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 48 kDa band observed in human muscle lysates (predicted MW of 50 kDa according to NP_060500). Recommended for use at 0.5-2 μg/mL.
ORP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ORP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: OSBPL1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
TILAUS
Laajenna kaikki / Pienennä kaikki
Pistosyvyys Pak. VWR-tuoteluettelon nro Määrä
100 µg 100 µG PRSI46-106
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
TEKNISET TIEDOT
Antigeenin symboli OSBPL1A
Antigeenin nimi Oxysterol Binding Protein-Like 1A
Reaktiivisuus Human
Konjugaatio Unconjugated
Klonaalisuus Polyclonal
Isäntä Goat
Western blot Yes
ELISA Yes
Antigen synonyms ORP1, ORP-1, OSBPL1B
Pitoisuus 500 ug/mL
Immunogeeni ORP1 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of ORP1.
Form Liquid
Syötä tarkemmat tiedot.
Valitse tuote johon etsit Sertifikaattia.
Ilmoita eränumero. Tämä kenttä on pakollinen. Jos et tiedä VWR Chemicalsin eränumeroa syötä "x".
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
tai
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Kuvaus Tuoteluettelon nro Availability Tuote Sinun hintasi Price Per lkm