Jatka istuntoa?
Jäljellä 
Tämä istunto vanhenee
Istuntosi on vanhentunut. Kirjasimme sinut ulos turvallisuussyistä.
Haluatko kirjautua uudelleen sisään?

Anti-OSBPL9 Goat Polyclonal Antibody

Toimittaja: ProSci Inc.
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 150 kDa and 19 kDa in lysates of human brain, and testis and 55 kDa in lysates of human placenta and Testes after 0.5 μg/mL antibody staining.5 kDa for NP_683702.1 and NP_683703.1; b: 63.9 kDa for NP_683704.1; c: 70.2 kDa for NP_683705.1; d: 81.8 kDa for NP_683706.1; e: 83.2 kDa for NP_078862.2; f: 84.3 kDa for NP_683707.1). All detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands). Immunohistochemistry: In paraffin embedded human lung shows staining of alveolar macrophages.. Recommended concentration, 2-4 μg/mL.

ORP9 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ORP9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: OSBPL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human




 

TILAUS

Laajenna kaikki / Pienennä kaikki
Pistosyvyys Pak. VWR-tuoteluettelon nro Määrä
100 µg 100 µG PRSI46-112
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

TEKNISET TIEDOT

Antigeenin symboli OSBPL9
Antigeenin nimi Oxysterol binding Protein-like 9
Reaktiivisuus Human
Konjugaatio Unconjugated
Klonaalisuus Polyclonal
Isäntä Goat
ELISA Yes
ImmunoChemistry Yes
Antigen synonyms ORP9, RP4-657D16.1, ORP-9, oxysterol-binding protein-related protein 9, OSBP-related protein 9
Pitoisuus 500 ug/mL
Immunogeeni ORP9 antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of ORP9.
Form Liquid
Syötä tarkemmat tiedot.

Valitse tuote johon etsit Sertifikaattia.

Ilmoita eränumero. Tämä kenttä on pakollinen. Jos et tiedä VWR Chemicalsin eränumeroa syötä "x".

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

tai
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Kuvaus Tuoteluettelon nro Availability Tuote Sinun hintasi Price Per lkm