Anti-PYGL Rabbit Polyclonal Antibody
Toimittaja: ProSci
PRSI48-157EA 480 EUR
PRSI48-157
Anti-PYGL Rabbit Polyclonal Antibody
PYGL antibody can be used in Western Blot starting at 1 μg/mL, and immunohistochemistry starting at 10 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PYGL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Gorilla, Human