Anti-GPR77 Rabbit Polyclonal Antibody
Toimittaja: ProSci
PRSI48-255EA 480 EUR
PRSI48-255
Anti-GPR77 Rabbit Polyclonal Antibody
C5L2 is an Anaphylatoxin Receptor. Binding to anaphylatoxins can result in increased triglyceride synthesis, although its precise role in cellular responses has not been determined.
GPR77 antibody can be used in immunohistochemistry starting at 10 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GPR77
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human