Human recombinant Lepr (from HEK293 cells)
Toimittaja: ProSci
PRSI96-513EA 584 EUR
PRSI96-513
Human recombinant Lepr (from HEK293 cells)
Proteiinit
Leptin receptor (LEPR) is also known as LEP-R, cluster of differentiation 295 (CD295), OB-R and B219, is a single-transmembrane-domain receptor of the gp130 family of cytokine receptors. Leptin receptor exists as homodimer and binds Leptin with high affinity, thus mediates the biological function of the adipocyte-specific hormone Leptin. LEPR is a receptor for leptin (an adipocyte-specific hormone that regulates body weight), and is involved in the regulation of fat metabolism, as well as in a novel hematopoietic pathway that is required for normal lymphopoiesis. Mutations in this protein have been associated with obesity and pituitary dysfunction. Interaction of leptin and leptin receptor is crucial for body weight and bone mass regulation in mammals through hypothalamic effects on satiety and energy expenditure. Meanwhile, research data supports a leptin receptor activation model based on ligand-induced conformational changes.