Whole blood, hepatitis C virus (HCV)
HCMSPB000F-2-HCVEA 0 EUR
HCMSPB000F-2-HCV
Whole blood, hepatitis C virus (HCV)
Whole blood used for researching the causes, diagnosis, supportive care and treatment of Hepatitis C Virus (HCV). The whole blood is collected from IRB-consented donors diagnosed with hepatitis C virus (HCV).
Storage: Use immediately upon receipt.
Varoitus: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.