Organic reference standard, Heparin sodium BRP, Ph.Eur., RS
Toimittaja: LGC Standards
EDQMH0200000EA 282 EUR
EDQMH0200000
Organic reference standard, Heparin sodium BRP, Ph.Eur., RS
Standardiaineet ja vertailumateriaalit Organic Standards
  • In Monographs 0569 and 20705
  • Solubility in water: 50 g/l at 20°C
  • Pharmaceutical applications