3-Methyl-5-isoxazolecarboxylic acid 97%

Varoitus

ACRO452660050EA 357 EUR
ACRO452660050
3-Methyl-5-isoxazolecarboxylic acid 97%
3-Methyl-5-isoxazolecarboxylic acid
Kaava: C₅H₅NO₃
MW: 127,10 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00464222
CAS-numero: 4857-42-5

Order Now

Testitulosten erittely

Appearance Off-white to brown Powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR