Cefaclor 96%
Toimittaja: Thermo Scientific
Vaara
ACRO455260010EA 72 EUR
ACRO455260010 ACRO455260050
Cefaclor 96%
Cefaclor
Kaava: C₁₅H₁₄ClN₃O₄S
MW: 385,8 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151471
CAS-numero: 53994-73-3
Order Now
Testitulosten erittely
Appearance White to light yellow Powder
Infrared spectrum Conforms
Water 3.0 to 9.0 %
HPLC 95.0 %