1,4-Phenylene diisocyanide 97%

Toimittaja: Thermo Scientific
Varoitus

ACRO457080010EA 348 EUR
ACRO457080010
1,4-Phenylene diisocyanide 97%
1,4-Phenylene diisocyanide
Kaava: C₈H₄N₂
MW: 128,13 g/mol
Sulamispiste: 165…166 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012383
CAS-numero: 935-16-0

Order Now

Testitulosten erittely

Appearance Off-white to brown Powder or crystals
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR