2,6-Dibromobenzoic acid 95%
|
Varoitus
|
ACRO458190050EA
263
EUR
ACRO458190050
2,6-Dibromobenzoic acid 95%
2,6-Dibromobenzoic acid
Kaava:
C₇H₄Br₂O₂ Sulamispiste: 141…142 °C Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00234254 CAS-numero: 601-84-3 |
Testitulosten erittely
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥94.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...