Robenidine hydrochloride 98%

Toimittaja: Thermo Scientific
Varoitus

ACRO459400250EA 237 EUR
ACRO459400250
Robenidine hydrochloride 98%
Robenidine hydrochloride
Kaava: C₁₅H₁₃Cl₂N₅·ClH
MW: 370,67 g/mol
Sulamispiste: 252…254 °C
Storage Temperature: Pakastin
MDL numero: MFCD01724404
CAS-numero: 25875-50-7

Order Now

Testitulosten erittely

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR