Sibutramine hydrochloride monohydrate

Varoitus

ACRO462200050EA 256 EUR
ACRO462200050 ACRO462200010
Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Kaava: C₁₇H₂₆ClN·HCl·H₂O
Sulamispiste: 196 °C
Storage Temperature: Pakastin
CAS-numero: 125494-59-9

Order Now

Testitulosten erittely

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 194 to 199 °C
Water 4.5 to 6 %
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR