High visibility waistcoats, flame retardant, Senra
Toimittaja: Sioen
SIOE494AA2MF0FCLEA 32.4 EUR
SIOE494AA2MF0FCL SIOE494AA2MF0FCXL SIOE494AA2MF0FC2XL SIOE494AA2MF0FC3XL SIOE494AA2MF0FCS SIOE494AA2MF0FCM
High visibility waistcoats, flame retardant, Senra
Työvaatteet
These flame retardant and anti-static waistcoats are made from 98% polyester flame retardant mesh + 2% AST.
  • V-neck
  • Closure with touch and close fastening
  • Flame retardant reflective tape (50 mm)
Back length: 70 cm (S - 3XL)
Weight: 120 g/m²