Measuring device for the fast evaluation of material moisture in firewood, timber, flake board, inlay, plaster, cement and lots more. A suitable characteristic is selected with help of material table on the rear side of the device before measuring. The measured value is displayed only a short time afterwards. The device is especially designed for precise firewood and timber measurements, however, a lot of additional building materials can be rated.
- Material tables on rear side of device
- Integrated and exchangeable measuring needles
- Moisture rating (wet/dry) via bar graph
- Display of material moisture or water content, integrated temperature compensation
Characteristic curves: Tree different wood groups (h.01, h.02, h.03) for a total of 130 wood types and eight different building material curves (c.01, c.02, c.03, c.04, c.05, c.06, c.07, c.08).
Sertifikaatit: DIN EN 13183