Goggles, Pilot
Toimittaja: Bollé Safety
111-0160EA 25.3 EUR
111-0160
Goggles, Pilot
Suojalasit, naamiomalliset
The goggles co-injected, ventilated frame guarantees unequalled adaptability and comfort.
  • Excellent field of vision
  • Very flexible PP/TPR sealed frame
  • Adjustable strap
  • Fits over prescription glasses
PLATINUM® coated PC lens: Coating on both sides ensures high scratch-resistance (1,4 cd/m²), high resistance to the most aggressive chemicals and slows the appearance of fogging.
Weight: 89 g