Silikageeli 30% in water, colloidal dispersion

43111.A7EA 190 EUR
43111.A7 43111.A1
Silikageeli 30% in water, colloidal dispersion
Silikageeli

As a binder in ceramic investment shell coating, with refractory powders for yielding superior face coats

Kaava: SiO₂
MW: 60,08 g/mol
Tiheys: 1,20
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011232
CAS-numero: 7631-86-9
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR