Lasihiili, spherical powder 1000-2000 µm Type 2

41531.18EA 349 EUR
41531.18
Lasihiili, spherical powder 1000-2000 µm Type 2
Lasihiili
Kaava: C
MW: 12,01 g/mol
Sulamispiste: 3652 °C
Tiheys: 1,95 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00133992
CAS-numero: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR