Volframi ≥99.96% (metals basis), sauvoja, Puratronic®, Ø 4.0 mm (0.16 in)

11446.HREA 847 EUR
11446.HR
Volframi ≥99.96% (metals basis), sauvoja, Puratronic®, Ø 4.0 mm (0.16 in)
Volframi
Kaava: W
MW: 183,84 g/mol
Kiehumispiste: 5660 °C (1013 hPa)
Sulamispiste: 3410 °C
Tiheys: 19 g/cm³ (20 °C)
MDL numero: MFCD00011461
CAS-numero: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR