2-bromi-2-metyylipropaani ≥98% stabiloitu
Toimittaja: Thermo Scientific
Vaara
Stabilization: Stabilised with potassium carbonate
A13727.36EA 432 EUR
A13727.36 A13727.14 A13727.22 ABCRAB114374.0100
2-bromi-2-metyylipropaani ≥98% stabiloitu
2-bromi-2-metyylipropaani
Purity: 98+%
CAS No.: 507-19-7
Molecular Formula: C4H9Br
Formula Weight: 137.02
Flash Point: 18°(64°F)
Physical Form: Liquid
Density: 1.210
Refractive Index: 1.4280
Appearance: Colourless
Melting Point: -20°
Boiling Point: 74-76°
Beilstein Reference: 1730892
Merck Reference: 14,1555
UN No.: 2342
MDL No.: MFCD00000125
Kaava: C₄H₉Br
MW: 137,02 g/mol
Kiehumispiste: 74…76 °C
Sulamispiste: –20 °C
Tiheys: 1,210
Leimahduspiste: 18 °C (64 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000125
CAS-numero: 507-19-7
EINECS: 208-065-9
UN: 2342
ADR: 3,II
Merck Index: 13,01554