Magnesiumkloridi 1 mol/l (2 N) vesiliuos for determining phosphatase in serum
220933MEA
110
EUR
220933M
Magnesiumkloridi 1 mol/l (2 N) vesiliuos for determining phosphatase in serum
Magnesiumkloridi
Kaava:
MgCl₂ MW: 95,21 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00011106 CAS-numero: 7786-30-3 EINECS: 232-094-6 Merck Index: 13,05684 |
Testitulosten erittely
Magnesium chloride | 0.999 - 1.001 mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...