Palladium 5% on barium sulphate hydrogenation catalyst, reduced

ACRO317530100EA 182 EUR
ACRO317530100
Palladium 5% on barium sulphate hydrogenation catalyst, reduced
Palladium
Kaava: Pd
MW: 106,420 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011167
CAS-numero: 7440-05-3
EINECS: 231-115-6

Order Now

Testitulosten erittely

Appearance Grey to brown to black Powder
Assay 4.0 to 6.0 % (Pd)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR