N-2-Aminoetyyli-1-naphftyyliamiinidihydrokloridi

Toimittaja: Thermo Scientific
Varoitus

ACRO20690-0250EA 0 EUR
ACRO20690-0250
N-2-Aminoetyyli-1-naphftyyliamiinidihydrokloridi
N-2-Aminoetyyli-1-naphftyyliamiinidihydrokloridi
Kaava: C₁₂H₁₆Cl₂N₂
MW: 259,18 g/mol
Sulamispiste: 188…190 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012556
CAS-numero: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://fi.vwr.com/stibo/web/std.lang.all/64/71/2346471.jpg

Order Now

Testitulosten erittely

Appearance White to beige to pink Powder
Infrared spectrum Conforms
Titration Total base ≥98.0 % (on dry substance)
Water ≤5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR