Lastat, Powder spatulas, disposable, SteriWare®
Toimittaja: Sampling Systems
SteriWare®
PP. These disposable spatulas feature a rounded handle and a robust blade.
  • Manufactured and packed in a controlled atmosphere (class 100000 cleanroom)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • Approved for use in ATEX Zones 0, 1 & 2 (contact Sampling Systems for full details)
  • Disposable: No cleaning is required before or after sampling
  • Available as gamma irradiated sterile (SAL 10-6)
Suitable for sterile sampling and dispensing quantities of powders in all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.).
TILAUS
Laajenna kaikki / Pienennä kaikki
Steriili Pituus Terän pituus Terän L Väri Pak. VWR-tuoteluettelon nro Määrä
- 230 mm 150 mm 19 mm sininen 100 SAMP8095B-150
+ 230 mm 150 mm 19 mm sininen 100 SAMP8095B-150S
- 330 mm 250 mm 19 mm sininen 100 SAMP8095B-250
+ 330 mm 250 mm 19 mm sininen 100 SAMP8095B-250S
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Syötä tarkemmat tiedot.
Valitse tuote johon etsit Sertifikaattia.
Ilmoita eränumero. Tämä kenttä on pakollinen. Jos et tiedä VWR Chemicalsin eränumeroa syötä "x".
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
tai
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Kuvaus Tuoteluettelon nro Availability Tuote Sinun hintasi Price Per lkm