Bottles, wide neck, with screw cap, round
Toimittaja: Rixius
RIXI10300100068ULMEA 3 EUR
RIXI10300100068ULM RIXI10301025055ULM RIXI10301010040ULM RIXI10300003032ULM RIXI10300005032ULM RIXI10300010040ULM RIXI10301100068ULM RIXI10300025055ULM RIXI10301003032ULM RIXI10301005032ULM RIXI10300050055ULM
Bottles, wide neck, with screw cap, round
Pullot Lasipullot
Clear and amber glass with thread and black PP cap.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Round thread in accordance with DIN 168
Ideal for sampling, storing and transporting powders and pastes.