1-Bromi-4-pentyylibentseeni

Toimittaja: Molekula

MOLE14094698-5GEA 0 EUR
MOLE14094698-5G
1-Bromi-4-pentyylibentseeni
1-Bromi-4-pentyylibentseeni
Kaava: C₁₁H₁₅Br
MW: 227,14 g/mol
MDL numero: MFCD00061113
CAS-numero: 51554-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR