ω,3-Dibromiasetofenoni

Toimittaja: Molekula

MOLEM66449379EA 65.5 EUR
MOLEM66449379
ω,3-Dibromiasetofenoni
ω,3-Dibromiasetofenoni
Kaava: C₈H₆Br₂O
MW: 277,94 g/mol
Sulamispiste: 51 °C
MDL numero: MFCD00082698
CAS-numero: 18523-22-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR