4-Chloro-3-nitrobenzyl alcohol

Toimittaja: Molekula

MOLE41754298-25GEA 0 EUR
MOLE41754298-25G
4-Chloro-3-nitrobenzyl alcohol
4-Chloro-3-nitrobenzyl alcohol
Kaava: C₇H₆ClNO₃
MW: 187,58 g/mol
Sulamispiste: 66 °C
MDL numero: MFCD00007086
CAS-numero: 55912-20-4
EINECS: 259-901-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR