Butyrylcholine chloride

Toimittaja: Molekula

MOLE49629139-25GEA 350 EUR
MOLE49629139-25G
Butyrylcholine chloride
Butyrylcholine chloride
Kaava: H₃C(CH₂)₂C(O)O(CH₂)₂N(CH₃)₃Cl
MW: 209,72 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD00011844
CAS-numero: 2963-78-2
EINECS: 220-999-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR