Ammoniumpersulfaatti (APS) >98%, PlusOne

Toimittaja: Cytiva
Vaara

Synonyms: Ammonium persulphate

17-1311-01EA 42.16 EUR
17-1311-01
Ammoniumpersulfaatti (APS) >98%, PlusOne
Ammoniumpersulfaatti (APS)

PlusOne™ ammonium persulfate is the initiator of choice for the polymerization of acrylamide. High quality of PlusOne™ ammonium Persulfate supports efficient polymerization at concentrations as low as 1 mM.


  • Used in IEF, PAGE and Sequencing
  • Soluble in cold water and hot water

Kaava: (NH₄)₂S₂O₈
MW: 228,2 g/mol
Sulamispiste: 120 °C (haj. lämpötila)
Tiheys: 1,98 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003390
CAS-numero: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://fi.vwr.com/stibo/web/std.lang.all/06/39/16640639.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR