Safety ankle boots, lace-up, Basic, Alex, Jori
Toimittaja: Elten
Alex Jori Basic Alex
ELTN16520/39EA 60.9 EUR
ELTN16520/39 ELTN16520/45 ELTN16520/44 ELTN16520/43 ELTN16520/42 ELTN16520/38 ELTN16520/48 ELTN16520/37 ELTN16520/36 ELTN16520/47 ELTN16520/46 ELTN16520/41 ELTN16520/40
Safety ankle boots, lace-up, Basic, Alex, Jori
Kengät ja saappaat
Durable safety ankle boots made from cowhide leather and hydrophobised textile Cordura® (tear and abrasion resistant, dirt and water repellent), reflective materials, skin-friendly textile lining, a full padded bellows tongue and collar padding and a dual-density PU/PU NEW BASIC sole (oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full-length inlay sole
  • Anti-static soft fleece insole
  • PU toe protection cap
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black/blue
Sertifikaatit: EN ISO 20345; S2, SRC