Anti-CALB2 Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-1800-50EA 302 EUR
BSENR-1800-50
Anti-CALB2 Rabbit Polyclonal Antibody
Vasta-aineet
Calretinin is a calcium-binding protein which is abundant in auditory neurons. Ref: uniprot.org. Antibody is specific for calretinin and does not recognize closely related proteins parvalbumin and calbindin as determined by Western Blotting.

  • Western blotting (1:5,000-1:10,000), Immunohistochemistry (1:5,000-1:10,000) and Immunocytochemistry (1:1,000-1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CALB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Cow
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR