Sterile C-Flex Adapters and Extensions are designed to facilitate and expedite the on-site construction of single-use fluid transfer assemblies.
- Manufactured in ISO Class 7 Cleanroom
- Supplied Gamma Irradiated to Sterility Assurance Level (SAL) 10-6
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, and production and cleanroom applications.
Assembled in an ISO Class 7 cleanroom and manufactured from USP Class VI materials. Tubing assemblies are offered with custom size and configuration capabilities. Tubing is weldable/sealable. All product contact materials are animal-free or processed in accordance to EMEA/410/01.
The assemblies are able to withstand temperatures from –20 to 65 °C (– 4 to 149 °F) without losing integrity. When used with sterile VWR® Bottle or Flask Assemblies, custom possibilities are endless.
Pakkaus: All assemblies are individually double-bagged with product label located on inner bag.