(4-Nitrophenyl)-β-D-glucopyranoside

Toimittaja: Thermo Scientific

J19594.06EA 892 EUR
J19594.06
(4-Nitrophenyl)-β-D-glucopyranoside
(4-Nitrophenyl)-β-D-glucopyranoside
Kaava: C₁₂H₁₅NO₈
MW: 301,25 g/mol
Sulamispiste: 166…167 °C
Storage Temperature: Pakastin
MDL numero: MFCD00006593
CAS-numero: 2492-87-7
EINECS: 219-661-3

Order Now

Testitulosten erittely

Assay (unspecified) - Absorbance (425 nm, 8 mg/ml, 1 cm) in 0.15 M AMP buffer, (pH 10.2) ≤0.07
Assay (unspecified) - Free p-Nitrophenol ≤0.01%
Identification (FTIR) Conforms
Water Content (Karl Fischer Titration) ≤10%
Optical Rotation (dry basis, c=1 in H₂O) -106 ±5°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR