Bovine Serum Albumin, Fraction V, heat-shock fractionated

Toimittaja: Thermo Fisher Scientific
J10857.18EA 176 EUR
J10857.18
Bovine Serum Albumin, Fraction V, heat-shock fractionated
Proteiinit
  • Protein: ≥96,0%
  • Residue on ignition: ≤2,0%
  • Loss on drying: ≤5,0%
  • pH (2%, H2O): 6,5 to 7,5
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR