Kolesteroli

J13580.22EA 1150 EUR
J13580.22
Kolesteroli
Kolesteroli
Kaava: C₂₇H₄₆O
MW: 386,66 g/mol
Kiehumispiste: 360 °C
Sulamispiste: 147…150 °C
Tiheys: 1,067
Storage Temperature: Jääkaappi
MDL numero: MFCD00003646
CAS-numero: 57-88-5
EINECS: 200-353-2

Order Now

Testitulosten erittely

Identification By IR
Thin Layer Chromatography Homogeneous

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR